Assay Details
| Gene Symbol: Oosp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133353.3 | NP_579931.1 | 4 - 5 | 508 | 71 | |
| GenBank mRNA | AB050982.2 | - | 4 - 5 | 501 | 71 | |
| AB086437.1 | - | 3 - 4 | 393 | 71 | ||
| AF420487.1 | - | 4 - 5 | 513 | 71 | ||
| AK033038.1 | - | 4 - 5 | 511 | 71 | ||
| AK162132.1 | - | 4 - 5 | 511 | 71 | ||
| BC100313.1 | - | 4 - 5 | 487 | 30936071 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | AK033038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | AK162132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | BC100313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | NM_133353.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | AB050982.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | AK033038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | AK162132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | BC100313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | NM_133353.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195135 | AB050982.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013431 | AK033038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013431 | AK162132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013431 | BC100313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013431 | NM_133353.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013431 | AB050982.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69135 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179877 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111491 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013431 | AK033038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013431 | AK162132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013431 | BC100313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013431 | NM_133353.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013431 | AB050982.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | AK033038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | AK162132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | BC100313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | NM_133353.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | AB050982.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | AK033038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | AK162132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | BC100313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | NM_133353.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195135 | AB050982.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957315 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03550889 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111491 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03550890 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179877 | - | - |
| Whole Mouse Genome G4122A | A_51_P205042 | AK033038.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205042 | AK162132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205042 | BC100313.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205042 | NM_133353.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205042 | AB050982.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | AK033038.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | AK162132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | BC100313.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | NM_133353.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | AB050982.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | AK033038.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | AK162132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | BC100313.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | NM_133353.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195135 | AB050982.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134435 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117985 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111491 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179877 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14485632 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14485629 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14277584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14485635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14189684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14302957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14302952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31957249 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10466247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466241 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466245 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418531_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10466239 | - | - |
| Mouse Genome 430 2.0 Array | 1418531_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418531_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170834_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170834_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900010719.mm.1 | NM_133353.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010719.mm.1 | NM_133353.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010719.mm.1 | TC1900001142.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900010720.mm.1 | NM_133353.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010720.mm.1 | NM_133353.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010720.mm.1 | TC1900001142.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900010718.mm.1 | NM_133353.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010718.mm.1 | NM_133353.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010718.mm.1 | TC1900001142.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418531_PM_at | - | - |
| Mouse Expression Set 430 | 1418531_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418531_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418531_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418531_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418531_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 428949 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3440008 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707996 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707996 | - | - |
| mouse-6 v1.1 (Illumina) | scl52718.5.1_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52718.5.1_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707996 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707996 | - | - |

Oosp1
Genomic Map