Assay Details
| Gene Symbol: Grid2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159321.1 | NP_001152793.1 | 16 - 17 | 3103 | 59 | |
| NM_133355.1 | NP_579933.1 | 15 - 16 | 2407 | 59 | ||
| GenBank mRNA | AF099933.2 | - | 15 - 16 | 2407 | 59 | |
| BC075624.1 | - | 2459 | 6841572 | 59 | ||
| DQ193535.1 | - | 16 - 17 | 3103 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grid2ip
Genomic Map