Assay Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133681.4 | NP_598442.1 | 2 - 3 | 200 | 97 | |
GenBank mRNA | AK078869.1 | - | 2 - 3 | 201 | 97 | |
AK162521.1 | - | 2 - 3 | 201 | 97 | ||
BC003448.1 | - | 2 - 3 | 140 | 2654254 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK078869.1,BC003448.1,NM_133681.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | AK162521.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | BC003448.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | AK078869.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | NM_133681.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | AK162521.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | BC003448.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | AK078869.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333923 | NM_133681.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379170 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935177 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129035 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179333 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | AK162521.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | BC003448.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | AK078869.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | NM_133681.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | AK162521.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | BC003448.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | AK078869.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333923 | NM_133681.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228090 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23228108 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179333 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05368194 | - | - |
Whole Mouse Genome G4122A | A_52_P61752 | AK162521.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P61752 | BC003448.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P61752 | AK078869.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P61752 | NM_133681.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P61752 | AK007428.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | AK162521.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | BC003448.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | AK078869.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | NM_133681.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | AK162521.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | BC003448.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | AK078869.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P333923 | NM_133681.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136345 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126351 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179333 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03690980 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03690992 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03690978 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03293210 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03690985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23227988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23228109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03955474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04050487 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03410234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04006071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03928702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03293210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03690978 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04050984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03690985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04051003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03690980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04051007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228097 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228074 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03690988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03690977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03955476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04051002 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03690976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03955472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03690982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23228103 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10515292 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515290 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515285 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515286 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515283 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515291 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515289 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515287 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515284 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515288 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417958_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417957_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10515282 | - | - |
Mouse Genome 430 2.0 Array | 1417958_at | - | - |
Mouse Genome 430 2.0 Array | 1417957_a_at | - | - |
Mouse Genome 430 2.0 Array | 1417958_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1417957_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66805_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66805_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400026604.mm.1 | TC0400003334.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400026604.mm.1 | NM_133681.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026604.mm.1 | NM_133681.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026599.mm.1 | TC0400003334.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400026599.mm.1 | NM_133681.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026599.mm.1 | NM_133681.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026601.mm.1 | TC0400003334.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400026601.mm.1 | NM_133681.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026601.mm.1 | NM_133681.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026603.mm.1 | TC0400003334.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400026603.mm.1 | NM_133681.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026603.mm.1 | NM_133681.4 | Refseq |
Mouse Expression Set 430 | 1417958_PM_at | - | - |
Mouse Expression Set 430 | 1417957_a_at | - | - |
Mouse Expression Set 430 | 1417958_at | - | - |
Mouse Expression Set 430 | 1417957_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1417957_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1417958_at | - | - |
Mouse Genome 430A 2.0 Array | 1417957_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1417958_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417958_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417957_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417958_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417957_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2450519 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2450519 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002450519 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624180 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2624180 | - | - |
mouse-6 v1.1 (Illumina) | scl23959.8_66-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2450519 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl23959.8_66-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2450519 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2624180 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2624180 | - | - |
