Assay Details
| Gene Symbol: Pycr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133705.2 | NP_598466.1 | 4 - 5 | 732 | 111 | |
| GenBank mRNA | AK007535.1 | - | 4 - 5 | 111 | ||
| AK150756.1 | - | 2 - 3 | 1852 | 111 | ||
| AK152302.1 | - | 2 - 3 | 1852 | 111 | ||
| AK165618.1 | - | 4 - 5 | 707 | 111 | ||
| AK167622.1 | - | 4 - 5 | 688 | 111 | ||
| BC006882.1 | - | 4 - 5 | 708 | 2647612 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pycr2
Genomic Map