Assay Details
| Gene Symbol: Clmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133733.4 | NP_598494.2 | 4 - 5 | 929 | 75 | |
| GenBank mRNA | AB040490.1 | - | 4 - 5 | 846 | 75 | |
| AK028983.1 | - | 4 - 5 | 899 | 75 | ||
| AK033723.1 | - | 4 - 5 | 931 | 75 | ||
| AK080481.1 | - | 4 - 5 | 884 | 75 | ||
| AY259213.1 | - | 5 - 6 | 558 | 75 | ||
| AY326421.1 | - | 4 - 5 | 564 | 75 | ||
| BC026447.1 | - | 4 - 5 | 755 | 4459717 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clmp
Genomic Map