Assay Details
| Gene Symbol: Ptcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133735.2 | NP_598496.3 | 5 - 6 | 1085 | 74 | |
| GenBank mRNA | AK088764.1 | - | 5 - 6 | 1004 | 74 | |
| AK143877.1 | - | 5 - 6 | 1042 | 74 | ||
| AK150514.1 | - | 5 - 6 | 1042 | 74 | ||
| AK151064.1 | - | 5 - 6 | 1053 | 74 | ||
| AK152077.1 | - | 5 - 6 | 1053 | 74 | ||
| AK153366.1 | - | 5 - 6 | 1053 | 74 | ||
| AK171484.1 | - | 5 - 6 | 1040 | 74 | ||
| BC004766.1 | - | 5 - 6 | 1003 | 3156304 | 74 | |
| BC019121.1 | - | 3 - 4 | 473 | 74 | ||
| BC083097.1 | - | 5 - 6 | 1044 | 30459747 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptcd1
Genomic Map