Assay Details
| Gene Symbol: Ptcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133735.2 | NP_598496.3 | 8 - 9 | 2083 | 77 | |
| GenBank mRNA | AK088764.1 | - | 8 - 9 | 2002 | 77 | |
| AK143877.1 | - | 8 - 9 | 2041 | 77 | ||
| AK150514.1 | - | 8 - 9 | 2040 | 77 | ||
| AK151064.1 | - | 8 - 9 | 2051 | 77 | ||
| AK152077.1 | - | 8 - 9 | 2051 | 77 | ||
| AK153366.1 | - | 8 - 9 | 2051 | 77 | ||
| AK171484.1 | - | 8 - 9 | 2038 | 77 | ||
| BC004766.1 | - | 8 - 9 | 2001 | 3156304 | 77 | |
| BC019121.1 | - | 6 - 7 | 1471 | 77 | ||
| BC083097.1 | - | 8 - 9 | 2042 | 30459747 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164621 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164620 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021545 | AK172986.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | BC083097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | BC004766.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | AK171484.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | AK166306.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | AK152077.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | BC083097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | BC004766.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | AK171484.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | AK166306.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163079 | AK152077.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634639 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169620 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82092 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126342 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154410 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | BC083097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | BC004766.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | AK171484.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | AK166306.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | AK152077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | BC083097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | BC004766.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | AK171484.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | AK166306.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163079 | AK152077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021545 | AK172986.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24164602 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126342 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01243938 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01243926 | - | - |
| Whole Mouse Genome G4122A | A_51_P163079 | BC083097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | BC004766.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | AK171484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | AK166306.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | AK152077.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | BC083097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | BC004766.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | AK171484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | AK166306.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163079 | AK152077.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P227065 | BC006861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P227065 | BC006861.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133320 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126342 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04923170 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04923171 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04139410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04923169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164620 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24164621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24164532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04985267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04923169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04591839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04680664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04680660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04923170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04200223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24164597 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10527454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535646 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535639 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416391_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425096_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535637 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10527448 | - | - |
| Mouse Genome 430 2.0 Array | 1416391_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425096_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416391_at | - | - |
| Mouse Genome 430 2.0 Array | 1425096_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71799_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71799_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032927.mm.1 | TC0500003590.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032927.mm.1 | NM_133735.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032927.mm.1 | NM_133735.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032930.mm.1 | TC0500003590.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032930.mm.1 | NM_133735.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032930.mm.1 | NM_133735.2 | Refseq |
| Mouse Expression Set 430 | 1416391_PM_at | - | - |
| Mouse Expression Set 430 | 1416391_at | - | - |
| Mouse Expression Set 430 | 1425096_a_at | - | - |
| Mouse Expression Set 430 | 1425096_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416391_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416391_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425096_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425096_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425096_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416391_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416391_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425096_a_at | - | - |

Ptcd1
Genomic Map