Assay Details
| Gene Symbol: Emc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133749.2 | NP_598510.1 | 3 - 4 | 532 | 129 | |
| GenBank mRNA | AJ250345.1 | - | 3 - 4 | 512 | 129 | |
| AJ278128.1 | - | 3 - 4 | 527 | 129 | ||
| AK013744.1 | - | 3 - 4 | 500 | 129 | ||
| AK088738.1 | - | 3 - 4 | 527 | 129 | ||
| AK211388.1 | - | 1 - 2 | 110 | 129 | ||
| BC008164.1 | - | 1 - 2 | 99 | 129 | ||
| BC080668.1 | - | 3 - 4 | 513 | 129 | ||
| BC115477.1 | - | 3 - 4 | 502 | 40100523 | 129 | |
| BC115478.1 | - | 3 - 4 | 502 | 40100524 | 129 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC008164.1 transcripts.
Set Membership: |
|

Emc7
Genomic Map