Assay Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133765.4 | NP_598526.3 | 3 - 4 | 493 | 67 | |
| GenBank mRNA | AB041586.1 | - | 3 - 4 | 342 | 67 | |
| AK140412.1 | - | 3 - 4 | 494 | 67 | ||
| AK163641.1 | - | 3 - 4 | 462 | 67 | ||
| BC026929.1 | - | 3 - 4 | 478 | 4194857 | 67 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB041586.1,BC026929.1 transcripts.
Set Membership: |
|

Fbxo31
Genomic Map