Assay Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133765.4 | NP_598526.3 | 5 - 6 | 737 | 104 | |
| GenBank mRNA | AB041586.1 | - | 5 - 6 | 586 | 104 | |
| AK009859.1 | - | 2 - 3 | 178 | 104 | ||
| AK140412.1 | - | 5 - 6 | 738 | 104 | ||
| AK163641.1 | - | 5 - 6 | 706 | 104 | ||
| BC026929.1 | - | 5 - 6 | 722 | 4194857 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo31
Genomic Map