Assay Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133765.4 | NP_598526.3 | 6 - 7 | 852 | 75 | |
| GenBank mRNA | AB041586.1 | - | 6 - 7 | 701 | 75 | |
| AK009859.1 | - | 3 - 4 | 293 | 75 | ||
| AK140412.1 | - | 6 - 7 | 853 | 75 | ||
| AK163641.1 | - | 6 - 7 | 821 | 75 | ||
| BC026929.1 | - | 6 - 7 | 837 | 4194857 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo31
Genomic Map