Assay Details
| Gene Symbol: Dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133803.2 | NP_598564.2 | 12 - 13 | 1456 | 126 | |
| GenBank mRNA | AK031020.1 | - | 12 - 13 | 1421 | 126 | |
| AK087994.1 | - | 12 - 13 | 1420 | 126 | ||
| AK149853.1 | - | 12 - 13 | 1421 | 126 | ||
| AK150101.1 | - | 12 - 13 | 1416 | 126 | ||
| AK150126.1 | - | 12 - 13 | 1410 | 126 | ||
| AK166842.1 | - | 12 - 13 | 1421 | 126 | ||
| AK168195.1 | - | 12 - 13 | 1409 | 126 | ||
| AK200538.1 | - | 2 - 3 | 110 | 126 | ||
| BC004600.1 | - | 11 - 12 | 1398 | 3584924 | 126 | |
| BC066817.1 | - | 12 - 13 | 1407 | 30429618 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp3
Genomic Map