Assay Details
Gene Symbol: AI597479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133818.1 | NP_598579.1 | 1 - 2 | 155 | 99 | |
GenBank mRNA | AK081372.1 | - | 1 - 2 | 146 | 99 | |
BC006931.1 | - | 1 - 2 | 155 | 3598975 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
