Assay Details
Gene Symbol: AI597479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133818.1 | NP_598579.1 | 3 - 4 | 694 | 59 | |
GenBank mRNA | AK081372.1 | - | 3 - 4 | 685 | 59 | |
BC006931.1 | - | 3 - 4 | 694 | 3598975 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20186992 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20186993 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20186991 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239292 | BC006931.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239292 | NM_133818.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239292 | AK050174.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239292 | BC006931.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239292 | NM_133818.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239292 | AK050174.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147815 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943953 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639227 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431813 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202233 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644883 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33273 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173041 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139697 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239292 | BC006931.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239292 | NM_133818.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239292 | AK050174.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239292 | BC006931.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239292 | NM_133818.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239292 | AK050174.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20186995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20186997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20186996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20186991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20186993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20187008 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173041 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00056576 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139697 | - | - |
Whole Mouse Genome G4122A | A_52_P628667 | AK081372.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P239292 | BC006931.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P239292 | NM_133818.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P239292 | AK050174.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P239292 | BC006931.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P239292 | NM_133818.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P239292 | AK050174.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118386 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139697 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173041 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00439008 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00070678 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00070679 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00439001 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20186992 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20186993 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20186991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20186991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20186989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20186993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20186997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20186995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20187026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00439017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187002 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20186991 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00439008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20186996 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00905228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20187007 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10345905 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345908 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345907 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345906 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417247_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10345904 | - | - |
Mouse Genome 430 2.0 Array | 1417247_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1417247_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98404_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98404_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100002381.mm.1 | NM_133818.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100002381.mm.1 | NM_133818.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100002381.mm.1 | TC0100000320.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100002382.mm.1 | NM_133818.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100002382.mm.1 | NM_133818.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100002382.mm.1 | TC0100000320.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417247_at | - | - |
Mouse Expression Set 430 | 1417247_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417247_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417247_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417247_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417247_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 667925 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2370408 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2370408 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002370408 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663092 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2663092 | - | - |
mouse-6 v1.1 (Illumina) | scl18013.4_456-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2370408 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl18013.4_456-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370408 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2970241 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2970241 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18013.4_456-S | - | - |
