Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133847.3 | NP_598608.2 | 15 - 16 | 1827 | 79 | |
| GenBank mRNA | AB093220.2 | - | 14 - 15 | 1571 | 79 | |
| AK035622.1 | - | 15 - 16 | 79 | |||
| AK048158.1 | - | 15 - 16 | 1585 | 79 | ||
| AK083541.1 | - | 15 - 16 | 1827 | 79 | ||
| AK087048.1 | - | 15 - 16 | 1580 | 79 | ||
| AK088012.1 | - | 15 - 16 | 1583 | 79 | ||
| AK162435.1 | - | 15 - 16 | 1588 | 79 | ||
| AK171487.1 | - | 15 - 16 | 1585 | 79 | ||
| BC006741.1 | - | 1 - 2 | 264 | 79 | ||
| BC071208.1 | - | 2 - 3 | 646 | 79 | ||
| BC110309.1 | - | 15 - 16 | 1583 | 5324218 | 79 | |
| BC116320.2 | - | 15 - 16 | 1779 | 40096002 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf4
Genomic Map