Assay Details
| Gene Symbol: Pafah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285872.1 | NP_001272801.1 | 8 - 9 | 913 | 109 | |
| NM_001285874.1 | NP_001272803.1 | 7 - 8 | 855 | 109 | ||
| NM_001285875.1 | NP_001272804.1 | 7 - 8 | 805 | 109 | ||
| NM_001285877.1 | NP_001272806.1 | 5 - 6 | 549 | 109 | ||
| NM_133880.3 | NP_598641.2 | 8 - 9 | 902 | 109 | ||
| GenBank mRNA | AK144090.1 | - | 8 - 9 | 865 | 109 | |
| AK145092.1 | - | 7 - 8 | 756 | 109 | ||
| AK156774.1 | - | 7 - 8 | 856 | 109 | ||
| AK181929.1 | - | 1 - 2 | 71 | 109 | ||
| BC021890.1 | - | 7 - 8 | 807 | 3598000 | 109 | |
| BC025495.1 | - | 8 - 9 | 924 | 5310200 | 109 | |
| BC025938.1 | - | 7 - 8 | 748 | 109 | ||
| BQ179290.1 | - | 6 - 7 | 624 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pafah2
Genomic Map