Assay Details
| Gene Symbol: Slc15a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133895.1 | NP_598656.1 | 4 - 5 | 1116 | 105 | |
| GenBank mRNA | AK048529.1 | - | 4 - 5 | 1083 | 105 | |
| AK154238.1 | - | 4 - 5 | 1085 | 105 | ||
| AK178039.1 | - | 2 - 3 | 149 | 105 | ||
| AK188139.1 | - | 2 - 3 | 149 | 105 | ||
| BC016233.1 | - | 4 - 5 | 1116 | 4502184 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a4
Genomic Map