Assay Details
| Gene Symbol: Papd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133905.2 | NP_598666.1 | 2 - 3 | 365 | 111 | |
| GenBank mRNA | AK146086.1 | - | 2 - 3 | 349 | 111 | |
| AM419011.1 | - | 1 - 2 | 138 | 111 | ||
| BC016629.1 | - | 2 - 3 | 365 | 3595512 | 111 | |
More Information
Set Membership: |
|

Papd4
Genomic Map