Assay Details
| Gene Symbol: Papd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133905.2 | NP_598666.1 | 5 - 6 | 806 | 82 | |
| GenBank mRNA | AK033141.1 | - | 2 - 3 | 143 | 82 | |
| AK146086.1 | - | 5 - 6 | 790 | 82 | ||
| AM419011.1 | - | 4 - 5 | 579 | 82 | ||
| BC016629.1 | - | 5 - 6 | 806 | 3595512 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033141.1 transcripts.
Set Membership: |
|

Papd4
Genomic Map