Assay Details
| Gene Symbol: Papd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133905.2 | NP_598666.1 | 11 - 12 | 1285 | 73 | |
| GenBank mRNA | AM419011.1 | - | 10 - 11 | 1046 | 73 | |
| BC016629.1 | - | 11 - 12 | 1285 | 3595512 | 73 | |
More Information
Set Membership: |
|

Papd4
Genomic Map