Assay Details
| Gene Symbol: Tbc1d14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113362.1 | NP_001106833.1 | 6 - 7 | 1283 | 67 | |
| NM_001113364.1 | NP_001106835.1 | 3 - 4 | 397 | 67 | ||
| NM_133910.3 | NP_598671.3 | 5 - 6 | 1102 | 67 | ||
| GenBank mRNA | AB064521.1 | - | 3 - 4 | 260 | 67 | |
| AB093293.1 | - | 4 - 5 | 896 | 67 | ||
| AK013092.1 | - | 3 - 4 | 323 | 67 | ||
| AK029986.1 | - | 6 - 7 | 1283 | 67 | ||
| BC011420.1 | - | 3 - 4 | 275 | 67 | ||
| BC042515.1 | - | 5 - 6 | 1078 | 4020510 | 67 | |
| BC086316.1 | - | 5 - 6 | 1170 | 30112465 | 67 | |
| BC096446.1 | - | 5 - 6 | 1094 | 6307706 | 67 | |
| BQ745653.1 | - | 3 - 4 | 402 | 67 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC011420.1 transcripts.
Set Membership: |
|

Tbc1d14
Genomic Map