Assay Details
| Gene Symbol: Tbc1d14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113362.1 | NP_001106833.1 | 10 - 11 | 1685 | 105 | |
| NM_001113364.1 | NP_001106835.1 | 7 - 8 | 799 | 105 | ||
| NM_133910.3 | NP_598671.3 | 9 - 10 | 1504 | 105 | ||
| GenBank mRNA | AB064521.1 | - | 7 - 8 | 105 | ||
| AB093293.1 | - | 8 - 9 | 1298 | 105 | ||
| AK029986.1 | - | 10 - 11 | 1685 | 105 | ||
| BC011420.1 | - | 7 - 8 | 677 | 105 | ||
| BC042515.1 | - | 9 - 10 | 1480 | 4020510 | 105 | |
| BC096446.1 | - | 9 - 10 | 1496 | 6307706 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d14
Genomic Map