Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 14 - 15 | 1718 | 89 | |
| NM_133914.3 | NP_598675.2 | 15 - 16 | 1856 | 89 | ||
| GenBank mRNA | AK014220.1 | - | 14 - 15 | 2005 | 89 | |
| AK044762.1 | - | 13 - 14 | 1801 | 89 | ||
| AK154335.1 | - | 15 - 16 | 1857 | 89 | ||
| AK155146.1 | - | 15 - 16 | 1857 | 89 | ||
| AK220218.1 | - | 15 - 16 | 1732 | 89 | ||
| AY591339.1 | - | 15 - 16 | 1757 | 89 | ||
| BC027230.1 | - | 14 - 15 | 1977 | 89 | ||
| BC057460.1 | - | 15 - 16 | 1710 | 4527934 | 89 | |
| DQ317932.1 | - | 14 - 15 | 1602 | 89 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map