Assay Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133922.3 | NP_598683.3 | 13 - 14 | 2188 | 114 | |
| GenBank mRNA | AB041543.1 | - | 11 - 12 | 1820 | 114 | |
| AK029235.1 | - | 4 - 5 | 634 | 114 | ||
| AK040525.1 | - | 13 - 14 | 2190 | 114 | ||
| AK128931.1 | - | 13 - 14 | 2139 | 114 | ||
| AK162934.1 | - | 13 - 14 | 1896 | 114 | ||
| AK173281.1 | - | 13 - 14 | 3892 | 114 | ||
| BC066816.1 | - | 13 - 14 | 2108 | 30084911 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krba1
Genomic Map