Assay Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133922.3 | NP_598683.3 | 14 - 15 | 2376 | 77 | |
| GenBank mRNA | AB041543.1 | - | 12 - 13 | 2008 | 77 | |
| AK029235.1 | - | 5 - 6 | 822 | 77 | ||
| AK040525.1 | - | 14 - 15 | 2378 | 77 | ||
| AK128931.1 | - | 14 - 15 | 2327 | 77 | ||
| AK162934.1 | - | 14 - 15 | 2084 | 77 | ||
| AK173281.1 | - | 14 - 15 | 4080 | 77 | ||
| AK196294.1 | - | 1 - 2 | 165 | 77 | ||
| BC066816.1 | - | 14 - 15 | 2296 | 30084911 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krba1
Genomic Map