Assay Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133922.3 | NP_598683.3 | 15 - 16 | 2514 | 102 | |
| GenBank mRNA | AB041543.1 | - | 13 - 14 | 2146 | 102 | |
| AK038090.1 | - | 1 - 2 | 164 | 102 | ||
| AK040525.1 | - | 15 - 16 | 2516 | 102 | ||
| AK128931.1 | - | 15 - 16 | 2465 | 102 | ||
| AK162934.1 | - | 15 - 16 | 2222 | 102 | ||
| AK173281.1 | - | 15 - 16 | 4218 | 102 | ||
| AK189155.1 | - | 1 - 2 | 102 | |||
| BC066816.1 | - | 15 - 16 | 2434 | 30084911 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krba1
Genomic Map