Assay Details
| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133933.4 | NP_598694.3 | 3 - 4 | 697 | 89 | |
| GenBank mRNA | AK029923.1 | - | 3 - 4 | 655 | 89 | |
| AK132750.1 | - | 3 - 4 | 670 | 89 | ||
| AK151899.1 | - | 3 - 4 | 661 | 89 | ||
| AK152003.1 | - | 3 - 4 | 670 | 89 | ||
| AK153867.1 | - | 3 - 4 | 656 | 89 | ||
| BC016080.1 | - | 3 - 4 | 659 | 4489287 | 89 | |
| BC085483.1 | - | 3 - 4 | 638 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpn1
Genomic Map