Assay Details
| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133933.4 | NP_598694.3 | 6 - 7 | 1196 | 104 | |
| GenBank mRNA | AK029923.1 | - | 6 - 7 | 1154 | 104 | |
| AK052130.1 | - | 6 - 7 | 1150 | 104 | ||
| AK132750.1 | - | 6 - 7 | 1169 | 104 | ||
| AK151899.1 | - | 6 - 7 | 1160 | 104 | ||
| AK152003.1 | - | 6 - 7 | 1169 | 104 | ||
| AK153867.1 | - | 6 - 7 | 1155 | 104 | ||
| BC002175.1 | - | 2 - 3 | 155 | 104 | ||
| BC016080.1 | - | 6 - 7 | 1158 | 4489287 | 104 | |
| BC085483.1 | - | 6 - 7 | 1137 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpn1
Genomic Map