Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600020729.mm.1 |
TC0600002666.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020729.mm.1 |
NM_133934.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020729.mm.1 |
NM_133934.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020731.mm.1 |
TC0600002666.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020731.mm.1 |
NM_133934.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020731.mm.1 |
NM_133934.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020732.mm.1 |
TC0600002666.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020732.mm.1 |
NM_133934.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020732.mm.1 |
NM_133934.4 |
Refseq |