Assay Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133934.4 | NP_598695.1 | 3 - 4 | 187 | 77 | |
| GenBank mRNA | AK017996.1 | - | 3 - 4 | 104 | 77 | |
| AK173116.1 | - | 155 | 77 | |||
| BC020103.1 | - | 3 - 4 | 151 | 3673446 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Isy1
Genomic Map