Assay Details
| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133945.1 | NP_598706.1 | 9 - 10 | 895 | 96 | |
| GenBank mRNA | AK150435.1 | - | 9 - 10 | 986 | 96 | |
| AK171673.1 | - | 9 - 10 | 900 | 96 | ||
| AK183463.1 | - | 2 - 3 | 96 | |||
| AK183527.1 | - | 4 - 5 | 278 | 96 | ||
| BC010473.1 | - | 8 - 9 | 878 | 3586304 | 96 | |
| BC024782.1 | - | 8 - 9 | 878 | 5123949 | 96 | |
| BC024839.1 | - | 9 - 10 | 895 | 5355521 | 96 | |
| BC027129.1 | - | 8 - 9 | 824 | 96 | ||
| BC034196.1 | - | 8 - 9 | 876 | 4985176 | 96 | |
| S71575.1 | - | 1 - 2 | 115 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vrk3
Genomic Map