Assay Details
| Gene Symbol: Kdelr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133950.2 | NP_598711.1 | 2 - 3 | 359 | 77 | |
| GenBank mRNA | AJ278132.1 | - | 2 - 3 | 359 | 77 | |
| AK013118.1 | - | 2 - 3 | 375 | 77 | ||
| AK078785.1 | - | 2 - 3 | 428 | 77 | ||
| AY027435.1 | - | 2 - 3 | 188 | 77 | ||
| BC011370.1 | - | 2 - 3 | 271 | 3152867 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdelr1
Genomic Map