Assay Details
| Gene Symbol: Cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133978.2 | NP_598739.1 | 2 - 3 | 502 | 63 | |
| GenBank mRNA | AF188504.1 | - | 2 - 3 | 338 | 63 | |
| AK151785.1 | - | 2 - 3 | 305 | 63 | ||
| AK167886.1 | - | 2 - 3 | 348 | 63 | ||
| AK168024.1 | - | 2 - 3 | 355 | 63 | ||
| AY241869.1 | - | 2 - 3 | 359 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm7
Genomic Map