Assay Details
| Gene Symbol: Upb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133995.4 | NP_598756.1 | 6 - 7 | 939 | 62 | |
| GenBank mRNA | AK149373.1 | - | 6 - 7 | 836 | 62 | |
| AK149480.1 | - | 6 - 7 | 836 | 62 | ||
| BC021388.1 | - | 6 - 7 | 824 | 5055165 | 62 | |
| BC024447.1 | - | 4 - 5 | 490 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Upb1
Genomic Map