Assay Details
| Gene Symbol: Fig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133999.1 | NP_598760.1 | 2 - 3 | 349 | 62 | |
| GenBank mRNA | AK041196.1 | - | 2 - 3 | 350 | 62 | |
| AK155392.1 | - | 2 - 3 | 346 | 62 | ||
| AK169881.1 | - | 2 - 3 | 313 | 62 | ||
| AK172130.1 | - | 2 - 3 | 343 | 62 | ||
| AK172341.1 | - | 2 - 3 | 313 | 62 | ||
| BC015295.1 | - | 2 - 3 | 349 | 4221630 | 62 | |
| BC031887.1 | - | 2 - 3 | 348 | 5363304 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fig4
Genomic Map