Assay Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199274.1 | NP_001186203.1 | 5 - 6 | 1057 | 66 | |
| NM_134017.2 | NP_598778.1 | 5 - 6 | 861 | 66 | ||
| GenBank mRNA | AB070267.1 | - | 5 - 6 | 828 | 66 | |
| AK075600.1 | - | 5 - 6 | 762 | 66 | ||
| AK075947.1 | - | 5 - 6 | 796 | 66 | ||
| AK153126.1 | - | 5 - 6 | 815 | 66 | ||
| AK162087.1 | - | 5 - 6 | 796 | 66 | ||
| BC003457.1 | - | 5 - 6 | 792 | 2648452 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mat2b
Genomic Map