Assay Details
| Gene Symbol: Vipas39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142580.1 | NP_001136052.1 | 4 - 5 | 465 | 87 | |
| NM_001142581.1 | NP_001136053.1 | 4 - 5 | 465 | 87 | ||
| NM_134044.3 | NP_598805.2 | 4 - 5 | 479 | 87 | ||
| GenBank mRNA | AK038165.1 | - | 4 - 5 | 433 | 87 | |
| AK046322.1 | - | 4 - 5 | 436 | 87 | ||
| AK049643.1 | - | 4 - 5 | 412 | 87 | ||
| AK079055.1 | - | 4 - 5 | 412 | 87 | ||
| AK082261.1 | - | 4 - 5 | 393 | 87 | ||
| AK145043.1 | - | 448 | 87 | |||
| BB610654.1 | - | 4 - 5 | 466 | 87 | ||
| BC016646.1 | - | 4 - 5 | 388 | 2651233 | 87 | |
| BC023716.1 | - | 4 - 5 | 490 | 5346489 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vipas39
Genomic Map