Assay Details
| Gene Symbol: Galnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081421.1 | NP_001074890.1 | 1 - 2 | 358 | 83 | |
| GenBank mRNA | AB045325.1 | - | 1 - 2 | 320 | 83 | |
| AB175683.1 | - | 1 - 2 | 179 | 83 | ||
| AK138883.1 | - | 1 - 2 | 313 | 83 | ||
| AK173105.1 | - | 2 - 3 | 470 | 83 | ||
| BC110634.1 | - | 2 - 3 | 453 | 40059147 | 83 | |
| BC125015.1 | - | 2 - 3 | 452 | 40059141 | 83 | |
More Information
Set Membership: |
|

Galnt16
Genomic Map