Assay Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134046.5 | NP_598807.2 | 3 - 4 | 840 | 74 | |
| GenBank mRNA | AK013198.1 | - | 3 - 4 | 512 | 74 | |
| AK018444.1 | - | 3 - 4 | 485 | 74 | ||
| AK029226.1 | - | 3 - 4 | 841 | 74 | ||
| AK134266.1 | - | 3 - 4 | 512 | 74 | ||
| BC023148.1 | - | 3 - 4 | 483 | 4011571 | 74 | |
| BC034399.1 | - | 3 - 4 | 499 | 4481047 | 74 | |
| BC036278.1 | - | 3 - 4 | 483 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpo
Genomic Map