Assay Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134046.5 | NP_598807.2 | 4 - 5 | 953 | 60 | |
| GenBank mRNA | AK013198.1 | - | 4 - 5 | 625 | 60 | |
| AK018444.1 | - | 4 - 5 | 598 | 60 | ||
| AK029226.1 | - | 4 - 5 | 954 | 60 | ||
| AK036089.1 | - | 3 - 4 | 458 | 60 | ||
| AK134266.1 | - | 4 - 5 | 625 | 60 | ||
| BC023148.1 | - | 4 - 5 | 596 | 4011571 | 60 | |
| BC034399.1 | - | 4 - 5 | 612 | 4481047 | 60 | |
| BC036278.1 | - | 4 - 5 | 596 | 60 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK036089.1 transcripts.
Set Membership: |
|

Cenpo
Genomic Map