Assay Details
| Gene Symbol: Dusp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037955.4 | NP_001033044.1 | 4 - 5 | 256 | 102 | |
| NM_134068.3 | NP_598829.1 | 4 - 5 | 256 | 102 | ||
| GenBank mRNA | AF237619.1 | - | 4 - 5 | 216 | 102 | |
| AK148708.1 | - | 4 - 5 | 224 | 102 | ||
| AK149363.1 | - | 4 - 5 | 215 | 102 | ||
| BC080718.1 | - | 4 - 5 | 206 | 102 | ||
| BC108362.1 | - | 4 - 5 | 206 | 4917645 | 102 | |
| CX236611.1 | - | 4 - 5 | 256 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dusp22
Genomic Map