Assay Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134115.2 | NP_598876.1 | 11 - 12 | 1306 | 73 | |
| GenBank mRNA | AY292399.1 | - | 11 - 12 | 1170 | 73 | |
| BC009658.1 | - | 11 - 12 | 1306 | 3484764 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stk38
Genomic Map