Assay Details
| Gene Symbol: Abhd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134130.1 | NP_598891.1 | 2 - 3 | 331 | 91 | |
| GenBank mRNA | AF429302.1 | - | 2 - 3 | 331 | 91 | |
| AK134676.1 | - | 2 - 3 | 347 | 91 | ||
| AK144238.1 | - | 2 - 3 | 351 | 91 | ||
| BC026770.1 | - | 2 - 3 | 331 | 4234600 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd3
Genomic Map