Assay Details
Gene Symbol: AI837181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256515.1 | NP_001243444.1 | 1 - 2 | 168 | 62 | |
GenBank mRNA | BC024516.1 | - | 1 - 2 | 155 | 4984074 | 62 |
CK622392.1 | - | 1 - 2 | 148 | 62 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC024516.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928610 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928608 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928607 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P244923 | BC024516.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P244923 | BC024516.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848023 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68447 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160988 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P244923 | BC024516.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P244923 | BC024516.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928629 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928608 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928613 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160988 | - | - |
Whole Mouse Genome G4122A | A_51_P244923 | BC024516.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P244923 | BC024516.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110530 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119257 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160988 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928607 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928608 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928624 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14303243 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14303246 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14325208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928625 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14144204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14303989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14402972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14192361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14303234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14303239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928654 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14303981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14349968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14349965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14202586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14263344 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14325204 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10460584 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10460583 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418775_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10460582 | - | - |
Mouse Genome 430 2.0 Array | 1418775_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418775_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107242_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107242_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900000805.mm.1 | NM_001256515.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000805.mm.1 | NM_001256515.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000805.mm.1 | NM_134149.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000805.mm.1 | NM_134149.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000805.mm.1 | TC1900000065.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900000804.mm.1 | NM_001256515.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000804.mm.1 | NM_001256515.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000804.mm.1 | NM_134149.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000804.mm.1 | NM_134149.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000804.mm.1 | TC1900000065.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418775_at | - | - |
Mouse Expression Set 430 | 1418775_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418775_at | - | - |
Mouse Genome 430A 2.0 Array | 1418775_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418775_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418775_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 353893 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2480273 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2480273 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002480273 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698902 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2698902 | - | - |
mouse-6 v1.1 (Illumina) | scl52839.2_29-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2480273 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52839.2_29-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480273 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2847950 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2847950 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl52839.2_29-S | - | - |
