Assay Details
| Gene Symbol: Acot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134188.3 | NP_598949.3 | 2 - 3 | 822 | 70 | |
| GenBank mRNA | AK141651.1 | - | 2 - 3 | 812 | 70 | |
| AK172617.1 | - | 2 - 3 | 818 | 70 | ||
| BC064469.1 | - | 2 - 3 | 793 | 6508650 | 70 | |
| Gene Symbol: Acot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012006.2 | NP_036136.1 | 2 - 3 | 666 | 70 | |
| GenBank mRNA | AK053680.1 | - | 679 | 70 | ||
| BC003911.1 | - | 2 - 3 | 663 | 70 | ||
| BC022951.1 | - | 1 - 2 | 1716 | 70 | ||
| BC028261.1 | - | 2 - 3 | 676 | 5100478 | 70 | |
| BC040749.1 | - | 1 - 2 | 1716 | 70 | ||
| Y14004.1 | - | 2 - 3 | 666 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
very long-chain fatty acid metabolic process
response to hypoxia
long-chain fatty acid metabolic process
acyl-CoA metabolic process
very long-chain fatty acid catabolic process
receptor binding
palmitoyl-CoA hydrolase activity
hydrolase activity
thiolester hydrolase activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
very long-chain fatty acid metabolic process
response to hypoxia
long-chain fatty acid metabolic process
acyl-CoA metabolic process
very long-chain fatty acid catabolic process
receptor binding
palmitoyl-CoA hydrolase activity
hydrolase activity
thiolester hydrolase activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595768 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595769 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595772 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P525183 | BC064469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P525183 | AK141651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P525183 | NM_134188.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P525183 | BC064469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P525183 | AK141651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P525183 | NM_134188.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702942 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128495 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P525183 | BC064469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P525183 | AK141651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P525183 | NM_134188.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P525183 | BC064469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P525183 | AK141651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P525183 | NM_134188.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28595764 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128495 | - | - |
| Whole Mouse Genome G4122A | A_52_P525183 | BC064469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P525183 | AK141651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P525183 | NM_134188.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P525183 | BC064469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P525183 | AK141651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P525183 | NM_134188.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126176 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128495 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09980273 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09980260 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09980270 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09980269 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09980265 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09980262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595768 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28595769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28595813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10197373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10184099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09980268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10287633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10122321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10197375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10356438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10129912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10009475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09895351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09895361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10182739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09895358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28595767 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10397146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10397147 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439478_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422996_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10397145 | - | - |
| Mouse Genome 430 2.0 Array | 1422996_at | - | - |
| Mouse Genome 430 2.0 Array | 1439478_at | - | - |
| Mouse Genome 430 2.0 Array | 1422996_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422997_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422997_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1439478_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171210_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171210_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200005681.mm.1 | NM_012006.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005681.mm.1 | NM_012006.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005681.mm.1 | TC1200000798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005680.mm.1 | NM_012006.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005680.mm.1 | NM_012006.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005680.mm.1 | TC1200000798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005676.mm.1 | NM_134188.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005676.mm.1 | NM_134188.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005676.mm.1 | TC1200000797.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005682.mm.1 | NM_012006.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005682.mm.1 | NM_012006.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005682.mm.1 | TC1200000798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005675.mm.1 | NM_134188.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005675.mm.1 | NM_134188.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005675.mm.1 | TC1200000797.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422996_PM_at | - | - |
| Mouse Expression Set 430 | 1422996_at | - | - |
| Mouse Expression Set 430 | 1422997_s_at | - | - |
| Mouse Expression Set 430 | 1422997_PM_s_at | - | - |
| Mouse Expression Set 430 | 1439478_PM_at | - | - |
| Mouse Expression Set 430 | 1439478_at | - | - |
| Murine Genome U74v2 Set | 103581_PM_at | - | - |
| Murine Genome U74v2 Set | 103581_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422997_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422997_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422996_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422996_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422996_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422996_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439478_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439478_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 480488 | - | - |
| ABI Mouse Genome Survey Microarray | 493258 | - | - |
| ABI Mouse Genome Survey Microarray | 663149 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4570021 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570021 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380722 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2745889 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243165 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2745889 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243165 | - | - |
| mouse-6 v1.1 (Illumina) | scl42992.3.1_66-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0171210.3_326-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380722 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570021 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0171210.3_326-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42992.3.1_66-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380722 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570021 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2745889 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2840492 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1243165 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2840492 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2745889 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243165 | - | - |

Acot2
Genomic Map