Assay Details
| Gene Symbol: Havcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134248.2 | NP_599009.2 | 4 - 5 | 629 | 84 | |
| GenBank mRNA | AF399829.1 | - | 4 - 5 | 620 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Havcr1
Genomic Map