Assay Details
| Gene Symbol: Elovl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134255.3 | NP_599016.2 | 3 - 4 | 417 | 83 | |
| GenBank mRNA | AK028761.1 | - | 3 - 4 | 357 | 83 | |
| AK031743.1 | - | 3 - 4 | 419 | 83 | ||
| AK045274.1 | - | 3 - 4 | 396 | 83 | ||
| AK051580.1 | - | 3 - 4 | 414 | 83 | ||
| AK085663.1 | - | 3 - 4 | 380 | 83 | ||
| AK085696.1 | - | 3 - 4 | 367 | 83 | ||
| AK088153.1 | - | 3 - 4 | 355 | 83 | ||
| AK162451.1 | - | 3 - 4 | 418 | 83 | ||
| AK167755.1 | - | 3 - 4 | 357 | 83 | ||
| AK168360.1 | - | 3 - 4 | 357 | 83 | ||
| AK168987.1 | - | 3 - 4 | 344 | 83 | ||
| BC022911.1 | - | 3 - 4 | 326 | 4457285 | 83 | |
| KF987081.1 | - | 3 - 4 | 300 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elovl5
Genomic Map