Assay Details
| Gene Symbol: Slc26a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134420.4 | NP_599252.2 | 2306 | 102 | ||
| GenBank mRNA | AB099881.1 | - | 1467 | 102 | ||
| AF248494.1 | - | 1652 | 102 | |||
| AK029238.1 | - | 3228 | 102 | |||
| AK046993.1 | - | 3566 | 102 | |||
| AK053088.1 | - | 1957 | 102 | |||
| AK080750.1 | - | 1611 | 102 | |||
| AK163670.1 | - | 2307 | 102 | |||
| AK163671.1 | - | 2307 | 102 | |||
| AY032863.1 | - | 1611 | 102 | |||
| AY049076.1 | - | 1566 | 102 | |||
| BC028856.1 | - | 1630 | 4165725 | 102 | ||
| CN692543.1 | - | 126 | 102 | |||
More Information
Set Membership: |
|

Slc26a6
Genomic Map