Assay Details
| Gene Symbol: Slc26a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134420.4 | NP_599252.2 | 2893 | 109 | ||
| GenBank mRNA | AB099881.1 | - | 2054 | 109 | ||
| AF248494.1 | - | 2239 | 109 | |||
| AK029238.1 | - | 3815 | 109 | |||
| AK046993.1 | - | 4153 | 109 | |||
| AY032863.1 | - | 2198 | 109 | |||
| AY049076.1 | - | 2153 | 109 | |||
| BC028856.1 | - | 2217 | 4165725 | 109 | ||
More Information
Set Membership: |
|

Slc26a6
Genomic Map