Assay Details
| Gene Symbol: Apobr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138310.1 | NP_612183.1 | 2 - 3 | 2496 | 111 | |
| GenBank mRNA | AF141335.1 | - | 2 - 3 | 2496 | 111 | |
| AK156422.1 | - | 2 - 3 | 2564 | 111 | ||
| BC030718.1 | - | 2 - 3 | 2504 | 4482457 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16842 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113780 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501244 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113780 | - | - |
| Whole Mouse Genome G4122A | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203182 | BC030718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203182 | AK156422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203182 | AF141335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203182 | NM_138310.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113534 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113780 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06116865 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06116887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06116860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06116839 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10557436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557437 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420382_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557434 | - | - |
| Mouse Genome 430 2.0 Array | 1420382_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420382_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171504_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171504_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700015408.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015408.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015408.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015401.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015401.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015401.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015403.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015403.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015403.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015407.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015407.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015407.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015404.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015404.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015404.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015409.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015409.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015409.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015402.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015402.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015402.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015405.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015405.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015405.mm.1 | NM_138310.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015406.mm.1 | TC0700001808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015406.mm.1 | NM_138310.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015406.mm.1 | NM_138310.1 | Refseq |
| Mouse Expression Set 430 | 1420382_at | - | - |
| Mouse Expression Set 430 | 1420382_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420382_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420382_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420382_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420382_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 930150 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2810070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2810070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002810070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2639734 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2639734 | - | - |
| mouse-6 v1.1 (Illumina) | scl32060.4.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2810070 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32060.4.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2639734 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2888834 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2888834 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2639734 | - | - |

Apobr
Genomic Map